Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methoxycarbonyl)phenyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 2-(Methoxycarbonyl)phenyl isothiocyanate
CAS 16024-82-1
Molecular Formula C9H7NO2S
Formula Weight 193.23
MDL Number MFCD00041062
UN Number UN2922
Beilstein 2720655
Synonym Methyl 2-isothiocyanatobenzoate; 2-Isothiocyanatobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2069306

 
alfa aesar™
B2069306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2069314

 
alfa aesar™
B2069314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.23
Chemical Name or Material 2-(Methoxycarbonyl)phenyl isothiocyanate
Assay 98%
Molecular Formula C9H7NO2S
MDL Number MFCD00041062
CAS 16024-82-1
UN Number UN2922
Beilstein 2720655
Synonym Methyl 2-isothiocyanatobenzoate; 2-Isothiocyanatobenzoic acid methyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Boiling Point 149° to 150°C (17mmHg)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.