Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methoxycarbonyl)phenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-(Methoxycarbonyl)phenyl isocyanate
CAS 1793-07-3
Molecular Formula C9H7NO3
Formula Weight 177.16g/mol
MDL Number MFCD00013848
UN Number UN2811
Beilstein 1241370
Synonym 2-(Carbomethoxy)phenyl isocyanate; Methyl 2-isocyanatobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1247103

 
alfa aesar™
L12471-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 45° to 49°C
Boiling Point 145° to 146°C (16mmHg)
Quantity 1g
Odor Irritating
Chemical Name or Material 2-(Methoxycarbonyl)phenyl isocyanate
CAS 1793-07-3
Assay 97%
Molecular Formula C9H7NO3
Formula Weight 177.16g/mol
MDL Number MFCD00013848
UN Number UN2811
Beilstein 1241370
Synonym 2-(Carbomethoxy)phenyl isocyanate; Methyl 2-isocyanatobenzoate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
Methyl 2-isocyanatobenzoate is used for the following syntheses 3H-quinazolin-4-ones, A1120, a high affinity (Ki = 8.3nM) non-retinoid ligand for the Retinol-binding protein, trans- and cis-N-phenyl-N′-(4-phenoxy)cyclohexylureas.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.

Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C