missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methoxycarbonyl)-6-nitrobenzoic Acid, TCI America™

$149.72 - $446.13

Chemical Identifiers

CAS 21606-04-2
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.156
MDL Number MFCD00034781
InChI Key DJMQLZPEBHSABD-UHFFFAOYSA-N
PubChem CID 4399477
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27955G View Documents TCI America
M27955G
5g Each for $149.72
 
M279525G View Documents TCI America
M279525G
25g Each for $446.13
 
Specifications

Chemical Identifiers

21606-04-2
225.156
DJMQLZPEBHSABD-UHFFFAOYSA-N
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O
C9H7NO6
MFCD00034781
4399477

Specifications

White-Yellow
5g
C9H7NO6
DJMQLZPEBHSABD-UHFFFAOYSA-N
225.156
225.16
≥98.0% (GC,T)
162°C
21606-04-2
MFCD00034781
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O
4399477
Crystal-Powder at 20°C
2-(Methoxycarbonyl)-6-nitrobenzoic Acid
Safety and Handling

Safety and Handling

TSCA : No