missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxycarbazole, TCI America™

$271.95 - $944.19

Chemical Identifiers

CAS 6933-49-9
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD09033506
InChI Key MDKVPSJRUXOFFV-UHFFFAOYSA-N
PubChem CID 10797986
IUPAC Name 2-methoxy-9H-carbazole
SMILES COC1=CC2=C(C=C1)C3=CC=CC=C3N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28171G View Documents TCI America
M28171G
1g Each for $271.95
 
M28175G View Documents TCI America
M28175G
5g Each for $944.19
 
Specifications

Chemical Identifiers

6933-49-9
197.237
MDKVPSJRUXOFFV-UHFFFAOYSA-N
2-methoxy-9H-carbazole
C13H11NO
MFCD09033506
10797986
COC1=CC2=C(C=C1)C3=CC=CC=C3N2

Specifications

White
1g
C13H11NO
MDKVPSJRUXOFFV-UHFFFAOYSA-N
2-methoxy-9H-carbazole
10797986
Crystal-Powder at 20°C
2-Methoxycarbazole
239°C
6933-49-9
MFCD09033506
COC1=CC2=C(C=C1)C3=CC=CC=C3N2
197.237
197.24
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No