Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxybiphenyl, 98+%

$74.51 - $74.51

Chemical Identifiers

Chemical Name or Material 2-Methoxybiphenyl
CAS 86-26-0
Molecular Formula C13H12O
Formula Weight 184.24
MDL Number MFCD00008367
Beilstein 2045749
Synonym 2-Phenylanisole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0106314

  SDS
alfa aesar™
L0106314
25g Each for $74.51
Description & Specifications

Specifications

Formula Weight 184.24
Chemical Name or Material 2-Methoxybiphenyl
TSCA TSCA
Assay ≥98%
Molecular Formula C13H12O
MDL Number MFCD00008367
CAS 86-26-0
Beilstein 2045749
Synonym 2-Phenylanisole
RTECS Number BZ8835000
Flash Point >110°C (230°F)
Boiling Point 273° to 274°C
Melting Point ≈30°C
Quantity 25g

Applications
2-Methoxybiphenyl is used as chemical and organic intermediates.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.