missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxybiphenyl 3-isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L0967406

 View more versions of this product

Catalog No. AAL0967406



Specifications

Specifications

2-Methoxybiphenyl 3-isothiocyanate
5g
C14H11NOS
UN2923
TZRMLJYZKZTEFW-UHFFFAOYSA-N
2-isothiocyanato-1-methoxy-4-phenylbenzene
617825
97%
78°C to 81°C
206761-68-4
MFCD00041078
2-methoxy-5-phenyl phenyl isothiocyanate, 2-methoxybiphenyl 3-isothiocyanate, 2-methoxy-5-phenyl phenylisothiocyanate, 3-isothiocyanato-4-methoxy-1,1'-biphenyl, 2-methoxy-5-phenylbenzenisothiocyanate, 3-isothiocyanato-4-methoxy-1,1'-biphenyl #, 1,1-biphenyl, 3-isothiocyanato-4-methoxy-9ci
COC1=C(C=C(C=C1)C2=CC=CC=C2)N=C=S
241.308
241.31

Chemical Identifiers

206761-68-4
241.308
TZRMLJYZKZTEFW-UHFFFAOYSA-N
617825
COC1=C(C=C(C=C1)C2=CC=CC=C2)N=C=S
C14H11NOS
MFCD00041078
2-methoxy-5-phenyl phenyl isothiocyanate, 2-methoxybiphenyl 3-isothiocyanate, 2-methoxy-5-phenyl phenylisothiocyanate, 3-isothiocyanato-4-methoxy-1,1'-biphenyl, 2-methoxy-5-phenylbenzenisothiocyanate, 3-isothiocyanato-4-methoxy-1,1'-biphenyl #, 1,1-biphenyl, 3-isothiocyanato-4-methoxy-9ci
2-isothiocyanato-1-methoxy-4-phenylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents