missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxybenzylamine, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC170542500

 View more versions of this product

Catalog No. AC170542500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methoxybenzylamine
Authentic
1.5475 to 1.5495 (20°C, 589nm)
250mL
97.5
98%
CH3OC6H4CH2NH2
13, 580
PXJACNDVRNAFHD-UHFFFAOYSA-N
(2-methoxyphenyl)methanamine
81292
Liquid
1.0500g/mL
Glass bottle
Colorless to Yellow
6850-57-3
100.0
C8H11NO
MFCD00008110
2-methoxybenzylamine, 2-methoxyphenyl methanamine, o-methoxybenzylamine, benzenemethanamine, 2-methoxy, 2-methoxy-benzylamine, 2-methoxyphenyl methylamine, 1-2-methoxyphenyl methanamine, 2-methyoxybenzylamine, 2-methoxy benzylamine, 2-methoxybenzyl amine
COC1=CC=CC=C1CN
137.182
137.18
≥97.5%

Chemical Identifiers

6850-57-3
137.182
PXJACNDVRNAFHD-UHFFFAOYSA-N
81292
COC1=CC=CC=C1CN
C8H11NO
MFCD00008110
2-methoxybenzylamine, 2-methoxyphenyl methanamine, o-methoxybenzylamine, benzenemethanamine, 2-methoxy, 2-methoxy-benzylamine, 2-methoxyphenyl methylamine, 1-2-methoxyphenyl methanamine, 2-methyoxybenzylamine, 2-methoxy benzylamine, 2-methoxybenzyl amine
(2-methoxyphenyl)methanamine
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

SDS Documents