Learn More
2-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2278406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methoxybenzoyl chloride | |
1.146 | |
84°C (184°F) | |
1.5724 | |
5 g | |
607605 | |
o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi | |
COC1=CC=CC=C1C(=O)Cl | |
170.592 | |
170.6 |
21615-34-9 | |
128°C to 129°C (8 mmHg) | |
C8H7ClO2 | |
MFCD00000664 | |
UN1729 | |
Moisture sensitive | |
RZNHSEZOLFEFGB-UHFFFAOYSA-N | |
2-methoxybenzoyl chloride | |
88969 | |
97% |
Chemical Identifiers
21615-34-9 | |
170.592 | |
RZNHSEZOLFEFGB-UHFFFAOYSA-N | |
88969 | |
COC1=CC=CC=C1C(=O)Cl |
C8H7ClO2 | |
MFCD00000664 | |
o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi | |
2-methoxybenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANISOYL CHLORIDE
EINECSNumber : 244-477-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only