missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methoxybenzonitrile 98.0+%, TCI America™
$68.02 - $209.03
Chemical Identifiers
CAS | 6609-56-9 |
---|---|
Molecular Formula | C8H7NO |
Molecular Weight (g/mol) | 133.15 |
MDL Number | MFCD00001783 |
InChI Key | FSTPMFASNVISBU-UHFFFAOYSA-N |
Synonym | 2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile |
PubChem CID | 81086 |
IUPAC Name | 2-methoxybenzonitrile |
SMILES | COC1=CC=CC=C1C#N |
Chemical Identifiers
6609-56-9 | |
133.15 | |
FSTPMFASNVISBU-UHFFFAOYSA-N | |
81086 | |
COC1=CC=CC=C1C#N |
C8H7NO | |
MFCD00001783 | |
2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile | |
2-methoxybenzonitrile |
Specifications
6609-56-9 | |
256°C | |
MFCD00001783 | |
2811 | |
FSTPMFASNVISBU-UHFFFAOYSA-N | |
2-methoxybenzonitrile | |
81086 | |
≥98.0% (GC) | |
2-Methoxybenzonitrile |
White-Yellow | |
C8H7NO | |
5 g | |
2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile | |
COC1=CC=CC=C1C#N | |
133.15 | |
133.15 | |
Crystalline Lumps |
Safety and Handling
TSCA : No