missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxybenzonitrile, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC433160050

 View more versions of this product

Catalog No. AC433160050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methoxybenzonitrile
Glass bottle
5g
98.5
99%
MFCD00001783
FSTPMFASNVISBU-UHFFFAOYSA-N
2-methoxybenzonitrile
81086
Liquid
Authentic
Colorless to Yellow
6609-56-9
100.0
C8H7NO
2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile
COC1=CC=CC=C1C#N
133.15
133.15
≥98.5%

Chemical Identifiers

6609-56-9
133.15
FSTPMFASNVISBU-UHFFFAOYSA-N
81086
COC1=CC=CC=C1C#N
C8H7NO
MFCD00001783
2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile
2-methoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents