Please login to your online account to display your discounted pricing

2-Methoxybenzenethiol, 97%, ACROS Organics™

$61.05 - $61.05

Chemical Identifiers

Chemical Name or Material 2-Methoxybenzenethiol
Name Note 97%
CAS 7217-59-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H8OS
Linear Formula CH3OC6H4SH
Formula Weight 140.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC179980010

  SDS
thermo fisher scientific
AC179980010
Glass bottle 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC179980500

  SDS
thermo fisher scientific
AC179980500
Glass bottle 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC179980050

  SDS
thermo fisher scientific
179980050
Glass bottle 5mL Each for $61.05
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 140.2
Quantity 1mL
Color Clear yellow
EINECS Number 230-605-7
TSCA TSCA
RTECS Number DC1790000
Density 1.14g/mL
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.5905 to 1.5925 (20°C, 589nm)
Synonym 2-Methoxythiophenol
Chemical Name or Material 2-Methoxybenzenethiol
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C7H8OS
Linear Formula CH3OC6H4SH
MDL Number MFCD00004834
Packaging Glass bottle
Health Hazard 1 STENCH
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 7217-59-6
Safety and Handling

STENCH

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.