missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-N-methylbenzylamine, 97%

Manufacturer:  Alfa Aesar™ H6333906

 View more versions of this product

Catalog No. AAH6333906



Specifications

Specifications

2-Methoxy-N-methylbenzylamine
1.5235
Colorless
6851-80-5
MFCD00025755
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, 2-methoxybenzyl methylamine, ksc495a5l
CNCC1=CC=CC=C1OC
151.209
151.21
97%
1.015g/mL
234°C to 235°C
5g
C9H13NO
UN2735
JCCQJCOMFAJJCQ-UHFFFAOYSA-N
1-(2-methoxyphenyl)-N-methylmethanamine
522302
Liquid

Chemical Identifiers

6851-80-5
151.209
JCCQJCOMFAJJCQ-UHFFFAOYSA-N
522302
CNCC1=CC=CC=C1OC
C9H13NO
MFCD00025755
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, 2-methoxybenzyl methylamine, ksc495a5l
1-(2-methoxyphenyl)-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

SDS Documents