missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-N-(4-methoxyphenyl)aniline, TCI America™

$262.78 - $785.30

Chemical Identifiers

CAS 58751-07-8
Molecular Formula C14H15NO2
MDL Number MFCD27956479
Synonym 2,4′-Dimethoxydiphenylamine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M30675G View Documents TCI America
M30675G
5g Each for $785.30
 
M30671G View Documents TCI America
M30671G
1g Each for $262.78
 
Specifications

Chemical Identifiers

58751-07-8
C14H15NO2
MFCD27956479
2,4′-Dimethoxydiphenylamine

Specifications

White-Yellow
5g
C14H15NO2
2,4′-Dimethoxydiphenylamine
Crystal-Powder at 20°C
2-Methoxy-N-(4-methoxyphenyl)aniline
71°C
58751-07-8
MFCD27956479
229.28
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No