missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-N-(3,4-dimethoxyphenyl)benzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 415696-09-2
Molecular Formula C16H17NO4
Molecular Weight (g/mol) 287.315
MDL Number MFCD00702339
InChI Key QLZIDCIZXPJKOS-UHFFFAOYSA-N
Synonym n-3,4-dimethoxyphenyl-2-methoxybenzamide, 2-methoxy-n-3,4-dimethoxyphenyl benzamide, cambridge id 5338121
PubChem CID 796052
IUPAC Name N-(3,4-dimethoxyphenyl)-2-methoxybenzamide
SMILES COC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59815MD View Documents Alfa Aesar™
H59815MD
250mg Each for $211.27
 
AAH5981503 View Documents Alfa Aesar™
H5981503
1g Each for $675.45
 
Specifications

Chemical Identifiers

415696-09-2
287.315
QLZIDCIZXPJKOS-UHFFFAOYSA-N
796052
COC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2OC)OC
C16H17NO4
MFCD00702339
n-3,4-dimethoxyphenyl-2-methoxybenzamide, 2-methoxy-n-3,4-dimethoxyphenyl benzamide, cambridge id 5338121
N-(3,4-dimethoxyphenyl)-2-methoxybenzamide

Specifications

250mg
C16H17NO4
n-3,4-dimethoxyphenyl-2-methoxybenzamide, 2-methoxy-n-3,4-dimethoxyphenyl benzamide, cambridge id 5338121
COC1=C(C=C(C=C1)NC(=O)C2=CC=CC=C2OC)OC
287.315
287.32
2-Methoxy-N-(3,4-dimethoxyphenyl)benzamide
415696-09-2
MFCD00702339
QLZIDCIZXPJKOS-UHFFFAOYSA-N
N-(3,4-dimethoxyphenyl)-2-methoxybenzamide
796052
97%