Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-6-(trifluoromethyl)phenylacetic acid, 97%

$112.70 - $376.69

Chemical Identifiers

Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)phenylacetic acid
CAS 1017779-10-0
Molecular Formula C10H9F3O3
Formula Weight 234.18
MDL Number MFCD09832328
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3209503

  SDS
alfa aesar™
H3209503
1g Each for $376.69

AAH32095MD

  SDS
alfa aesar™
H32095MD
250mg Each for $112.70
Description & Specifications

Specifications

Formula Weight 234.18
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)phenylacetic acid
CAS 1017779-10-0
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Molecular Formula C10H9F3O3
MDL Number MFCD09832328
Melting Point 148° to 150°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.