Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-6-(trifluoromethyl)benzoic acid, 97%

$160.08 - $628.03

Chemical Identifiers

Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzoic acid
CAS 119692-41-0
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD09832319
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3278903

  SDS
alfa aesar™
H3278903
1g Each for $160.08

AAH3278906

  SDS
alfa aesar™
H3278906
5g Each for $628.03
Description & Specifications

Specifications

Formula Weight 220.15
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C9H7F3O3
MDL Number MFCD09832319
CAS 119692-41-0
Melting Point 131° to 134°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.