Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-6-(trifluoromethyl)benzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzaldehyde
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD09832324
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3220603

 
alfa aesar™
H3220603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3220606

 
alfa aesar™
H3220606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.15
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C9H7F3O2
MDL Number MFCD09832324
Melting Point 46° to 49°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
Used as catalyst in diethylzinc addition to aldehydes.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.