Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-6-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)aniline
Molecular Formula C8H8F3NO
Formula Weight 191.15
MDL Number MFCD09832384
UN Number UN2810
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3244903

  SDS
alfa aesar™
H3244903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3244906

  SDS
alfa aesar™
H3244906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.15
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)aniline
Assay 97%
Molecular Formula C8H8F3NO
MDL Number MFCD09832384
UN Number UN2810
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.