missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-6-methylpyridine, TCI America™

$35.14 - $103.89

Chemical Identifiers

CAS 63071-03-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
InChI Key WIMNZMOEBDPZTB-UHFFFAOYSA-N
Synonym 2-methoxy-6-methyl-pyridine, 2-methoxy-6-picoline, 6-methoxy-2-picoline, 2-methoxy-6-methylam, acmc-209yyt, 6-methoxy-2-methylpyridine, 2-methoxy-6-methyl pyridine, ksc490s3b, 2-methyl-6-methyloxy pyridine, 2-methoxy-6-methylpyridine 1g
PubChem CID 5324773
IUPAC Name 2-methoxy-6-methylpyridine
SMILES CC1=NC(=CC=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M169525G View Documents TCI America
M169525G
25g Each for $103.89
 
M16955G View Documents TCI America
M16955G
5g Each for $35.14
 
Specifications

Chemical Identifiers

63071-03-4
123.155
2-methoxy-6-methyl-pyridine, 2-methoxy-6-picoline, 6-methoxy-2-picoline, 2-methoxy-6-methylam, acmc-209yyt, 6-methoxy-2-methylpyridine, 2-methoxy-6-methyl pyridine, ksc490s3b, 2-methyl-6-methyloxy pyridine, 2-methoxy-6-methylpyridine 1g
2-methoxy-6-methylpyridine
C7H9NO
WIMNZMOEBDPZTB-UHFFFAOYSA-N
5324773
CC1=NC(=CC=C1)OC

Specifications

156°C
25g
C7H9NO
2-methoxy-6-methyl-pyridine, 2-methoxy-6-picoline, 6-methoxy-2-picoline, 2-methoxy-6-methylam, acmc-209yyt, 6-methoxy-2-methylpyridine, 2-methoxy-6-methyl pyridine, ksc490s3b, 2-methyl-6-methyloxy pyridine, 2-methoxy-6-methylpyridine 1g
CC1=NC(=CC=C1)OC
123.155
123.16
≥98.0% (GC,T)
Yellow
63071-03-4
1993
WIMNZMOEBDPZTB-UHFFFAOYSA-N
2-methoxy-6-methylpyridine
5324773
Clear Liquid at 20°C
2-Methoxy-6-methylpyridine
Safety and Handling

Safety and Handling

TSCA : No