Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-6-methylpyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Methoxy-6-methylpyridine
CAS 63071-03-4
Molecular Formula C7H9NO
Formula Weight 123.16
MDL Number MFCD04112507
UN Number UN1993
Synonym 6-Methoxy-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2577203

 
alfa aesar™
H2577203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2577206

 
alfa aesar™
H2577206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 123.16
Chemical Name or Material 2-Methoxy-6-methylpyridine
Assay 98%
Molecular Formula C7H9NO
MDL Number MFCD04112507
CAS 63071-03-4
UN Number UN1993
Synonym 6-Methoxy-2-picoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H302-H312-H332-H315-H319
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 49°C (120°F)
Boiling Point 156°C
Quantity 1g

Applications
2-Methoxy-6-methylpyridine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H302-H312-H332-H315-H319
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.