Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-6-(methylamino)pyridine, 98%

$76.50 - $253.99

Chemical Identifiers

Chemical Name or Material 2-Methoxy-6-(methylamino)pyridine
CAS 88569-83-9
Molecular Formula C7H10N2O
Formula Weight 138.17
MDL Number MFCD00191557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2004806

  SDS
alfa aesar™
L2004806
5g Each for $76.50

AAL2004814

  SDS
alfa aesar™
L2004814
25g Each for $253.99
Description & Specifications

Specifications

Formula Weight 138.17
Chemical Name or Material 2-Methoxy-6-(methylamino)pyridine
Assay 98%
Molecular Formula C7H10N2O
MDL Number MFCD00191557
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 88569-83-9
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.