Please login to your online account to display your discounted pricing

2-Methoxy-5-(trifluoromethyl)benzoic acid, 97% Alfa Aesar™

Used as an intermediate in synthetic chemistry.

$97.12 - $289.38

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)benzoic acid
CAS 4864-01-1
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD01862091
Additional Information Incompatible with strong oxidizing agents, bases and strong acids
Solubility Information Insoluble in water
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2636203

 
alfa aesar
H2636203
1g Each for $97.12

AAH2636206

 
alfa aesar
H2636206
5g Each for $289.38
Description & Specifications

Specifications

Formula Weight 220.15
Additional Information Incompatible with strong oxidizing agents, bases and strong acids
Solubility Information Insoluble in water
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C9H7F3O3
MDL Number MFCD01862091
CAS 4864-01-1
Melting Point 108° to 111°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Methoxy-5-(trifluoromethyl)benzoic acid is used as an intermediate in synthetic chemistry.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, bases and strong acids.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.