Please login to your online account to display your discounted pricing

2-Methoxy-5-(trifluoromethyl)aniline, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)aniline
CAS 349-65-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H8F3NO
Linear Formula CH3OC6H3(CF3)NH2
Formula Weight 191.15
MDL Number MFCD00042486
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC306000050

  SDS
thermo fisher scientific
306000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 191.15
Quantity 5g
Color Beige-gray to brown
Melting Point 57° to 60°C
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C8H8F3NO
MDL Number MFCD00042486
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Packaging Glass bottle
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)aniline
Linear Formula CH3OC6H3(CF3)NH2
Synonym 3-Amino-4-methoxybenzotrifluoride
CAS 349-65-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.