Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Methoxy-5'-(trifluoromethyl)acetophenone, 97%

$180.81 - $718.73

Chemical Identifiers

Chemical Name or Material 2'-Methoxy-5'-(trifluoromethyl)acetophenone
CAS 503464-99-1
Molecular Formula C10H9F3O2
Formula Weight 218.17
MDL Number MFCD09832300
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3265203

  SDS
alfa aesar™
H3265203
1g Each for $180.81

AAH3265206

  SDS
alfa aesar™
H3265206
5g Each for $718.73
Description & Specifications

Specifications

Formula Weight 218.17
Chemical Name or Material 2'-Methoxy-5'-(trifluoromethyl)acetophenone
Assay 97%
Molecular Formula C10H9F3O2
MDL Number MFCD09832300
CAS 503464-99-1
Melting Point 45° to 47°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
In synthesis and evaluation of disubstituted N1- and N3-Imidazolidin-2-ones Acting as Potential Immunosuppressive Agents

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.