missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-5-(trifluoromethoxy)benzaldehyde 98.0+%, TCI America™

$144.84 - $434.52

Chemical Identifiers

CAS 145742-65-0
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD04115958
InChI Key ATRDCTRZAJKDPL-UHFFFAOYSA-N
Synonym 2-formyl-4-trifluoromethoxy anisole, 2-methoxy-5-trifluoromethoxy benzaldehyde, 2-methoxy-5-trifluoromethoxy-benzaldehyde, 2-methoxy-5-trifluoromethoxybenzaidehyde, 2-methoxy-5-trifluoromethyloxy benzaldehyde, acmc-209cvk, benzaldehyde, 2-methoxy-5-trifluoromethoxy, benzaldehyde,2-methoxy-5-trifluoromethoxy
PubChem CID 2783364
IUPAC Name 2-methoxy-5-(trifluoromethoxy)benzaldehyde
SMILES COC1=C(C=C(C=C1)OC(F)(F)F)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19455G View Documents Tci America
M19455G
5g Each for $144.84
 
M194525G View Documents Tci America
M194525G
25g Each for $434.52
 
Specifications

Chemical Identifiers

145742-65-0
220.147
ATRDCTRZAJKDPL-UHFFFAOYSA-N
2783364
COC1=C(C=C(C=C1)OC(F)(F)F)C=O
C9H7F3O3
MFCD04115958
2-formyl-4-trifluoromethoxy anisole, 2-methoxy-5-trifluoromethoxy benzaldehyde, 2-methoxy-5-trifluoromethoxy-benzaldehyde, 2-methoxy-5-trifluoromethoxybenzaidehyde, 2-methoxy-5-trifluoromethyloxy benzaldehyde, acmc-209cvk, benzaldehyde, 2-methoxy-5-trifluoromethoxy, benzaldehyde,2-methoxy-5-trifluoromethoxy
2-methoxy-5-(trifluoromethoxy)benzaldehyde

Specifications

Yellow
145742-65-0
MFCD04115958
ATRDCTRZAJKDPL-UHFFFAOYSA-N
2-methoxy-5-(trifluoromethoxy)benzaldehyde
2783364
Clear Liquid at 20°C
2-Methoxy-5-(trifluoromethoxy)benzaldehyde
5g
C9H7F3O3
2-formyl-4-trifluoromethoxy anisole, 2-methoxy-5-trifluoromethoxy benzaldehyde, 2-methoxy-5-trifluoromethoxy-benzaldehyde, 2-methoxy-5-trifluoromethoxybenzaidehyde, 2-methoxy-5-trifluoromethyloxy benzaldehyde, acmc-209cvk, benzaldehyde, 2-methoxy-5-trifluoromethoxy, benzaldehyde,2-methoxy-5-trifluoromethoxy
COC1=C(C=C(C=C1)OC(F)(F)F)C=O
220.147
220.15
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No