Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-5-(trifluoromethoxy)benzaldehyde, 98%

$48.66 - $116.77

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-(trifluoromethoxy)benzaldehyde
CAS 145742-65-0
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD04115958
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2610503

  SDS
alfa aesar™
H2610503
1g Each for $116.77

AAH26105MD

  SDS
alfa aesar™
H26105MD
250mg Each for $48.66
Description & Specifications

Specifications

Formula Weight 220.15
Chemical Name or Material 2-Methoxy-5-(trifluoromethoxy)benzaldehyde
Assay 98%
Molecular Formula C9H7F3O3
MDL Number MFCD04115958
CAS 145742-65-0
Melting Point 20° to 25°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.