Please login to your online account to display your discounted pricing

2-Methoxy-5-nitrophenol, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC397540050

 View more versions of this product

Catalog No. AC397540050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 169.14
Quantity 5g
EINECS Number 211-269-0
Linear Formula CH3OC6H3(NO2)OH
CAS 636-93-1
Chemical Name or Material 2-Methoxy-5-nitrophenol
Assay Percent Range 98%
Molecular Formula C7H7NO4
MDL Number MFCD00015561

Specifications

Chemical Name or Material 2-Methoxy-5-nitrophenol
Assay Percent Range 98%
Molecular Formula C7H7NO4
Linear Formula CH3OC6H3(NO2)OH
Formula Weight 169.14
MDL Number MFCD00015561