Please login to your online account to display your discounted pricing

2-Methoxy-5-nitrophenol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-nitrophenol
Assay Percent Range 98%
Molecular Formula C7H7NO4
Linear Formula CH3OC6H3(NO2)OH
Formula Weight 169.14
MDL Number MFCD00015561
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC397540050

  SDS
thermo fisher scientific
AC397540050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC397540250

  SDS
thermo fisher scientific
AC397540250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.14
Quantity 5g
EINECS Number 211-269-0
Linear Formula CH3OC6H3(NO2)OH
CAS 636-93-1
Chemical Name or Material 2-Methoxy-5-nitrophenol
Assay Percent Range 98%
Molecular Formula C7H7NO4
MDL Number MFCD00015561