missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-5-nitrophenol, 98%

Manufacturer:  Alfa Aesar™ L1182218

 View more versions of this product

Catalog No. AAL1182218



Specifications

Specifications

2-Methoxy-5-nitrophenol
103°C to 106°C
636-93-1
MFCD00015561
5-nitroguaiacol, phenol, 2-methoxy-5-nitro, 3-hydroxy-4-methoxynitrobenzene, 2-methoxy-5-nitro-phenol, unii-e5t11i2jub, e5t11i2jub, 2-hydroxy-4-nitroanisole, acmc-1b3lw, 2-methoxy-5-nitro phenol, 2-methyloxy-5-nitrophenol
COC1=C(C=C(C=C1)[N+](=O)[O-])O
169.136
CHEBI:81049
98%
110°C to 112°C (1mmHg)
50g
C7H7NO4
2047074
KXKCTSZYNCDFFG-UHFFFAOYSA-N
2-methoxy-5-nitrophenol
69471
169.14

Chemical Identifiers

636-93-1
169.136
KXKCTSZYNCDFFG-UHFFFAOYSA-N
69471
2-methoxy-5-nitrophenol
C7H7NO4
MFCD00015561
5-nitroguaiacol, phenol, 2-methoxy-5-nitro, 3-hydroxy-4-methoxynitrobenzene, 2-methoxy-5-nitro-phenol, unii-e5t11i2jub, e5t11i2jub, 2-hydroxy-4-nitroanisole, acmc-1b3lw, 2-methoxy-5-nitro phenol, 2-methyloxy-5-nitrophenol
CHEBI:81049
COC1=C(C=C(C=C1)[N+](=O)[O-])O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents