Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-5-nitrophenol, 98%

$61.72 - $626.49

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-nitrophenol
CAS 636-93-1
Molecular Formula C7H7NO4
Formula Weight 169.14
MDL Number MFCD00015561
Beilstein 2047074
Synonym 3-Hydroxy-4-methoxynitrobenzene; 5-Nitroguaiacol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1182209

  SDS
alfa aesar™
L1182209
10g Each for $61.72

AAL1182218

  SDS
alfa aesar™
L1182218
50g Each for $200.11

AAL1182230

  SDS
alfa aesar™
L1182230
250g Each for $626.49
Description & Specifications

Specifications

Formula Weight 169.14
Chemical Name or Material 2-Methoxy-5-nitrophenol
Assay 98%
Molecular Formula C7H7NO4
MDL Number MFCD00015561
Boiling Point 110° to 112°C (1mmHg)
CAS 636-93-1
Synonym 3-Hydroxy-4-methoxynitrobenzene; 5-Nitroguaiacol
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2047074
Melting Point 103° to 106°C
Quantity 10g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.