Please login to your online account to display your discounted pricing

2-Methoxy-5-nitroaniline, 99%, Acros Organics


Quantity 50g

Manufacturer: thermo fisher scientific  171300500

 View more versions of this product

Catalog No. AC171300500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Amorphous Powder
Assay ≥98.5%
Color Orange to brownish
Formula Weight 168.15
Quantity 50g
Beilstein 13, 389
RTECS Number BZ7175000
Melting Point 116° to 120°C
TSCA TSCA
EINECS Number 202-770-5
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
CAS Min % 98.5
Synonym 5-Nitro-o-anisidine
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 99-59-2
Name Note 99%
Assay Percent Range 99%
Linear Formula CH3OC6H3(NO2)NH2
Chemical Name or Material 2-Methoxy-5-nitroaniline
Molecular Formula C7H8N2O3
MDL Number MFCD00007261

Specifications

Chemical Name or Material 2-Methoxy-5-nitroaniline
Name Note 99%
CAS 99-59-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O3
Linear Formula CH3OC6H3(NO2)NH2
Formula Weight 168.15
MDL Number MFCD00007261
Synonym 5-Nitro-o-anisidine
Accessories

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents