Please login to your online account to display your discounted pricing

2-Methoxy-5-nitroaniline, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-nitroaniline
Name Note 99%
CAS 99-59-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2O3
Linear Formula CH3OC6H3(NO2)NH2
Formula Weight 168.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC171300500

  SDS
thermo fisher scientific
171300500
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Amorphous Powder
Assay ≥98.5%
Formula Weight 168.15
Color Orange to brownish
Quantity 50g
Beilstein 13, 389
EINECS Number 202-770-5
TSCA TSCA
RTECS Number BZ7175000
Melting Point 116° to 120°C
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 5-Nitro-o-anisidine
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Chemical Name or Material 2-Methoxy-5-nitroaniline
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H8N2O3
Linear Formula CH3OC6H3(NO2)NH2
MDL Number MFCD00007261
CAS 99-59-2
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.