Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-5-nitroaniline, 98+%

$38.49 - $252.45

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-nitroaniline
CAS 99-59-2
Molecular Formula C7H8N2O3
Formula Weight 168.15
MDL Number MFCD00007261
Beilstein 879620
Synonym 2-Amino-4-nitroanisole; 5-Nitro-o-anisidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2559114

 
alfa aesar™
B2559114
25g Each for $38.49

AAB2559122

 
alfa aesar™
B2559122
100g Each for $84.04

AAB2559136

 
alfa aesar™
B2559136
500g Each for $252.45
Description & Specifications

Specifications

Formula Weight 168.15
Chemical Name or Material 2-Methoxy-5-nitroaniline
TSCA TSCA
Assay ≥98%
Molecular Formula C7H8N2O3
MDL Number MFCD00007261
CAS 99-59-2
Beilstein 879620
Melting Point 117° to 119°C
Synonym 2-Amino-4-nitroanisole; 5-Nitro-o-anisidine
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H351-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BZ7175000
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H351-H332
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if inhaled.