missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-5-methylphenyl isothiocyanate, 99%

Manufacturer:  Alfa Aesar™ L1108506

 View more versions of this product

Catalog No. AAL1108506



Specifications

Specifications

2-Methoxy-5-methylphenyl isothiocyanate
288°C
190774-56-2
MFCD00041083
6919550
CYKFAZDMLSXWKZ-UHFFFAOYSA-N
2-isothiocyanato-1-methoxy-4-methylbenzene
601672
99%
>110°C (230°F)
5g
C9H9NOS
UN2922
2-methoxy-5-methylphenyl isothiocyanate, 2-methoxy-5-methylphenylisothiocyanate, benzene,2-isothiocyanato-1-methoxy-4-methyl, acmc-20anrv, 2-methoxy-5-methylbenzenisothiocyanate, 2-isothiocyanato-1-methoxy-4-methyl-benzene, #, 2-methoxy-5-methylphenyl isothiocyanate, benzene, 2-isothiocyanato-1-methoxy-4-methyl-9ci
CC1=CC(=C(C=C1)OC)N=C=S
179.237
179.24

Chemical Identifiers

190774-56-2
179.237
CYKFAZDMLSXWKZ-UHFFFAOYSA-N
601672
CC1=CC(=C(C=C1)OC)N=C=S
C9H9NOS
MFCD00041083
2-methoxy-5-methylphenyl isothiocyanate, 2-methoxy-5-methylphenylisothiocyanate, benzene,2-isothiocyanato-1-methoxy-4-methyl, acmc-20anrv, 2-methoxy-5-methylbenzenisothiocyanate, 2-isothiocyanato-1-methoxy-4-methyl-benzene, #, 2-methoxy-5-methylphenyl isothiocyanate, benzene, 2-isothiocyanato-1-methoxy-4-methyl-9ci
2-isothiocyanato-1-methoxy-4-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents