missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-5-methylphenyl isocyanate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441770010

 View more versions of this product

Catalog No. AC441770010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Methoxy-5-methylphenyl isocyanate
110°C
Glass Bottle
240.0°C
25.0°C to 26.0°C
59741-04-7
100.0
C9H9NO2
IDOHLSFNBKNRRJ-UHFFFAOYSA-N
2-isocyanato-1-methoxy-4-methylbenzene
4141520
Liquid or Low Melting Solid
1.1120g/mL
Authentic
1.112
Colorless
1g
97.5
97.5% min. (GC)
2-isocyanato-1-methoxy-4-methyl-benzene, 2-isocyanato-1-methyloxy-4-methylbenzene, 2-methoxy-5-methylbenzenisocyanate, 2-methoxy-5-methylpheny lisocyanate, 2-methoxy-5-methylphenyl isocyanate, 2-methoxy-5-methylphenylisocyanate, acmc-20anrw, benzene, 2-isocyanato-1-methoxy-4-methyl, benzene, 2-isocyanato-1-methoxy-4-methyl-9ci
CC1=CC(=C(C=C1)OC)N=C=O
163.18
163.18
98%

Chemical Identifiers

59741-04-7
163.18
2-isocyanato-1-methoxy-4-methyl-benzene, 2-isocyanato-1-methyloxy-4-methylbenzene, 2-methoxy-5-methylbenzenisocyanate, 2-methoxy-5-methylpheny lisocyanate, 2-methoxy-5-methylphenyl isocyanate, 2-methoxy-5-methylphenylisocyanate, acmc-20anrw, benzene, 2-isocyanato-1-methoxy-4-methyl, benzene, 2-isocyanato-1-methoxy-4-methyl-9ci
2-isocyanato-1-methoxy-4-methylbenzene
C9H9NO2
IDOHLSFNBKNRRJ-UHFFFAOYSA-N
4141520
CC1=CC(=C(C=C1)OC)N=C=O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reactio

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Danger

SDS Documents