Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-5-methylphenol, 98%

$67.58 - $237.05

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-methylphenol
CAS 1195-09-1
Molecular Formula C8H10O2
Formula Weight 138.17
MDL Number MFCD00040901
Beilstein 1817644
Synonym Isocreosol; 5-Methylguaiacol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0904404

 
alfa aesar™
L0904404
2g Each for $67.58

AAL0904409

 
alfa aesar™
L0904409
10g Each for $237.05
Description & Specifications

Specifications

Formula Weight 138.17
CAS 1195-09-1
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Methoxy-5-methylphenol
MDL Number MFCD00040901
Beilstein 1817644
Synonym Isocreosol; 5-Methylguaiacol
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 115° to 117°C (20mmHg)
Quantity 2g
Assay 98%
Molecular Formula C8H10O2
RTECS Number GP1770000
Melting Point 35° to 39°C

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.