missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-5-methylbenzonitrile, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438630010

 View more versions of this product

Catalog No. AC438630010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methoxy-5-methylbenzonitrile
Authentic
90°C (1.0mmHg)
1g
96.0
96% min. (GC)
2-methoxy-5-methyl-benzonitrile, 2-methoxy-5-methylbenzonitrile, acmc-1an2t, benzonitrile,2-methoxy-5-methyl
CC1=CC(=C(C=C1)OC)C#N
147.18
147.18
97%
>110°C
Glass bottle
Colorless to Yellow
53078-70-9
100.0
C9H9NO
MYOASWSDRGHGLN-UHFFFAOYSA-N
2-methoxy-5-methylbenzonitrile
3801023
Liquid

Chemical Identifiers

53078-70-9
147.18
2-methoxy-5-methyl-benzonitrile, 2-methoxy-5-methylbenzonitrile, acmc-1an2t, benzonitrile,2-methoxy-5-methyl
2-methoxy-5-methylbenzonitrile
C9H9NO
MYOASWSDRGHGLN-UHFFFAOYSA-N
3801023
CC1=CC(=C(C=C1)OC)C#N
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS Documents