missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methoxy-5-methylbenzonitrile, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438630050
Specifications
2-Methoxy-5-methylbenzonitrile | |
Authentic | |
90°C (1.0mmHg) | |
5g | |
96.0 | |
96% min. (GC) | |
2-methoxy-5-methyl-benzonitrile, 2-methoxy-5-methylbenzonitrile, acmc-1an2t, benzonitrile,2-methoxy-5-methyl | |
CC1=CC(=C(C=C1)OC)C#N | |
147.18 | |
147.18 | |
97% |
>110°C | |
Glass bottle | |
Colorless to Yellow | |
53078-70-9 | |
100.0 | |
C9H9NO | |
MYOASWSDRGHGLN-UHFFFAOYSA-N | |
2-methoxy-5-methylbenzonitrile | |
3801023 | |
Liquid |
Chemical Identifiers
53078-70-9 | |
147.18 | |
2-methoxy-5-methyl-benzonitrile, 2-methoxy-5-methylbenzonitrile, acmc-1an2t, benzonitrile,2-methoxy-5-methyl | |
2-methoxy-5-methylbenzonitrile |
C9H9NO | |
MYOASWSDRGHGLN-UHFFFAOYSA-N | |
3801023 | |
CC1=CC(=C(C=C1)OC)C#N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning