Please login to your online account to display your discounted pricing

2-Methoxy-5-formylphenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methoxy-5-formylphenylboronic acid
CAS 127972-02-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9BO4
Formula Weight 179.97
MDL Number MFCD01319044
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC359542500

  SDS
thermo fisher scientific
35954-2500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 179.97
Physical Form Powder
Quantity 250mg
Color Light orange
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C8H9BO4
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD01319044
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2-Methoxy-5-formylphenylboronic acid
CAS 127972-02-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.