Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-4-vinylphenol, 97%, stab. with 0.01% BHT

$144.80 - $516.82

Chemical Identifiers

Chemical Name or Material 2-Methoxy-4-vinylphenol
CAS 7786-61-0
Name Note stabilized with 0.01% BHT
Molecular Formula C9H10O2
Formula Weight 150.18
MDL Number MFCD00015437
Beilstein 2044521
Synonym 4-Hydroxy-3-methoxystyrene; 4-Vinylguaiacol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1319403

  SDS
alfa aesar™
A1319403
1g Each for $144.80

AAA1319406

  SDS
alfa aesar™
A1319406
5g Each for $516.82
Description & Specifications

Specifications

Formula Weight 150.18
Chemical Name or Material 2-Methoxy-4-vinylphenol
TSCA TSCA
Name Note stabilized with 0.01% BHT
Assay 97%
Molecular Formula C9H10O2
MDL Number MFCD00015437
Boiling Point 108° to 110°C (3mmHg)
CAS 7786-61-0
Beilstein 2044521
Melting Point 25° to 29°C
Synonym 4-Hydroxy-3-methoxystyrene; 4-Vinylguaiacol
Quantity 1g
Flash Point >100°C (212°F)
Odor Pleasant
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Recommended Storage 2° to 8°C
RTECS Number SL8205000

Applications
2-Methoxy-4-vinylphenol is used as a flavoring agent. It is also used as an aromatic substance resembling buckwheat. Further, it is used for chemical signaling in insects such as Red palm weevil. It is also utilized in imitation vanilla, coffee and cocoa.

Solubility
Miscible with water.

Notes
Air, light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C