Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-4-(trifluoromethyl)benzoic acid, 97%

$93.43 - $93.43

Chemical Identifiers

Chemical Name or Material 2-Methoxy-4-(trifluoromethyl)benzoic acid
CAS 448-36-2
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD04115976
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2637003

 
alfa aesar™
H2637003
1g Each for $93.43
Description & Specifications

Specifications

Formula Weight 220.15
Chemical Name or Material 2-Methoxy-4-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C9H7F3O3
MDL Number MFCD04115976
CAS 448-36-2
Melting Point 125° to 130°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Methoxy-4-(trifluoromethyl)benzoic acid is used as a synthetic building block.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.