Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-4-(trifluoromethyl)benzaldehyde, 97%

$93.76 - $831.22

Chemical Identifiers

Chemical Name or Material 2-Methoxy-4-(trifluoromethyl)benzaldehyde
CAS 132927-09-4
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD04115975
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2679703

 
alfa aesar™
H2679703
1g Each for $93.76

AAH2679706

 
alfa aesar™
H2679706
5g Each for $277.07

AAH2679714

 
alfa aesar™
H2679714
25g Each for $831.22
Description & Specifications

Specifications

Formula Weight 204.15
Assay 97%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Methoxy-4-(trifluoromethyl)benzaldehyde
Molecular Formula C9H7F3O2
MDL Number MFCD04115975
Melting Point 51° to 55°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 132927-09-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.