Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-4-(trifluoromethoxy)benzoic acid, 97%

$82.26 - $455.04

Chemical Identifiers

Chemical Name or Material 2-Methoxy-4-(trifluoromethoxy)benzoic acid
CAS 848947-91-1
Molecular Formula C9H7F3O4
Formula Weight 236.15
MDL Number MFCD04115966
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2653803

  SDS
alfa aesar™
H2653803
1g Each for $82.26

AAH2653809

  SDS
alfa aesar™
H2653809
10g Each for $455.04
Description & Specifications

Specifications

Formula Weight 236.15
Quantity 1g
MDL Number MFCD04115966
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H7F3O4
Chemical Name or Material 2-Methoxy-4-(trifluoromethoxy)benzoic acid
CAS 848947-91-1
Assay 97%
Melting Point 87° to 89°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.