Please login to your online account to display your discounted pricing

2-Methoxy-4-pyridinecarboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methoxy-4-pyridinecarboxylic acid
CAS 105596-63-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7NO3
Formula Weight 153.14
Synonym 2-Methoxyisonicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442410010

 
thermo fisher scientific
AC442410010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442410050

 
thermo fisher scientific
AC442410050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solid or Crystalline Powder
Assay ≥96.0%
Formula Weight 153.14
Quantity 1g
Color White to pale brown
CAS Min % 96.0
CAS Max % 100.0
Synonym 2-Methoxyisonicotinic acid
Chemical Name or Material 2-Methoxy-4-pyridinecarboxylic acid
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H7NO3
CAS 105596-63-2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.