Please login to your online account to display your discounted pricing

2-Methoxy-4-methylphenol, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  413851000

 View more versions of this product

Catalog No. AC413851000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Color Clear yellow, may darken during storage
Formula Weight 138.17
Physical Form Liquid
Quantity 100g
TSCA TSCA
EINECS Number 202-252-9
RTECS Number GP1755000
Density 1.0900g/mL
Merck Index 15, 2559
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5340 to 1.5380 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Synonym Creosol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 99%
Chemical Name or Material 2-Methoxy-4-methylphenol
MDL Number MFCD00002378
Name Note 99%
Molecular Formula C8H10O2
CAS 93-51-6

Specifications

Chemical Name or Material 2-Methoxy-4-methylphenol
Name Note 99%
CAS 93-51-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C8H10O2
Formula Weight 138.17
MDL Number MFCD00002378
Synonym Creosol
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents