missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-4-methyl-5-nitropyridine, TCI America™

$73.33 - $252.09

Chemical Identifiers

CAS 6635-90-1
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.152
MDL Number MFCD03095075
InChI Key DJNQRLCFAHKFLZ-UHFFFAOYSA-N
Synonym 2-methoxy-5-nitro-4-picoline, 2-methoxy-5-nitro-4-icoline, 2-methoxy-4-methyl-5-nitro-pyridine, pyridine, 2-methoxy-4-methyl-5-nitro, 2-methoxy-5-nitro-4-methylpyridine, 2-methoxy-5-nitro-4-picoline, pubchem6607, acmc-20ans4, ksc358g6j, 2-methoxy-4-methyl-5-nitropyridine
PubChem CID 243169
IUPAC Name 2-methoxy-4-methyl-5-nitropyridine
SMILES CC1=CC(=NC=C1[N+](=O)[O-])OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26521G View Documents TCI America
M26521G
1g Each for $73.33
 
M26525G View Documents TCI America
M26525G
5g Each for $252.09
 
Specifications

Chemical Identifiers

6635-90-1
168.152
DJNQRLCFAHKFLZ-UHFFFAOYSA-N
243169
CC1=CC(=NC=C1[N+](=O)[O-])OC
C7H8N2O3
MFCD03095075
2-methoxy-5-nitro-4-picoline, 2-methoxy-5-nitro-4-icoline, 2-methoxy-4-methyl-5-nitro-pyridine, pyridine, 2-methoxy-4-methyl-5-nitro, 2-methoxy-5-nitro-4-methylpyridine, 2-methoxy-5-nitro-4-picoline, pubchem6607, acmc-20ans4, ksc358g6j, 2-methoxy-4-methyl-5-nitropyridine
2-methoxy-4-methyl-5-nitropyridine

Specifications

White-Yellow
1g
C7H8N2O3
2-methoxy-5-nitro-4-picoline, 2-methoxy-5-nitro-4-icoline, 2-methoxy-4-methyl-5-nitro-pyridine, pyridine, 2-methoxy-4-methyl-5-nitro, 2-methoxy-5-nitro-4-methylpyridine, 2-methoxy-5-nitro-4-picoline, pubchem6607, acmc-20ans4, ksc358g6j, 2-methoxy-4-methyl-5-nitropyridine
CC1=CC(=NC=C1[N+](=O)[O-])OC
168.152
168.15
≥98.0% (GC)
81°C
6635-90-1
MFCD03095075
DJNQRLCFAHKFLZ-UHFFFAOYSA-N
2-methoxy-4-methyl-5-nitropyridine
243169
Crystal-Powder at 20°C
2-Methoxy-4-methyl-5-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No