Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-4-methyl-5-nitropyridine, 95%

Chemical Identifiers

Chemical Name or Material 2-Methoxy-4-methyl-5-nitropyridine
CAS 6635-90-1
Formula Weight 168.15
MDL Number MFCD03095075
Synonym 2-Methoxy-5-nitro-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6125803

  SDS
alfa aesar™
H6125803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6125806

  SDS
alfa aesar™
H6125806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.15
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Methoxy-4-methyl-5-nitropyridine
Physical Form Solid
Assay 95%
MDL Number MFCD03095075
CAS 6635-90-1
Synonym 2-Methoxy-5-nitro-4-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Quantity 1g

Applications
2-Methoxy-4-methyl-5-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.