missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC367540050

 View more versions of this product

Catalog No. AC367540050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
250.11
White to cream
100.0
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester
97%
Glass bottle
C13H19BO4
97%
Crystalline Powder
5g
101° to 106°C
96.0
2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
269410-22-2
Authentic
MFCD02093723

Chemical Identifiers

269410-22-2
100.0
C13H19BO4
MFCD02093723
2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
96.0
97%
250.11
4-Hydroxy-3-methoxyphenylboronic acid pinacol ester
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

Recommended Storage : Refrigerator +4°C