Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-3-(trifluoromethyl)phenylacetic acid, 97%

Chemical Identifiers

Chemical Name or Material 2-Methoxy-3-(trifluoromethyl)phenylacetic acid
Molecular Formula C10H9F3O3
Formula Weight 234.18
MDL Number MFCD09832317
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3167103

  SDS
alfa aesar™
H3167103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH31671MD

  SDS
alfa aesar™
H31671MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.18
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H9F3O3
Quantity 1g
Melting Point 71° to 75°C
MDL Number MFCD09832317
Chemical Name or Material 2-Methoxy-3-(trifluoromethyl)phenylacetic acid
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.