Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methoxy-3-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Methoxy-3-(trifluoromethyl)aniline
CAS 634187-08-9
Molecular Formula C8H8F3NO
Formula Weight 191.15
MDL Number MFCD08532489
UN Number UN2810
Synonym 3-Amino-2-methoxybenzotrifluoride; 3-Trifluoromethyl-o-anisidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3204503

 
alfa aesar™
H3204503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3204506

 
alfa aesar™
H3204506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.15
Chemical Name or Material 2-Methoxy-3-(trifluoromethyl)aniline
Assay 97%
Molecular Formula C8H8F3NO
MDL Number MFCD08532489
UN Number UN2810
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
CAS 634187-08-9
Synonym 3-Amino-2-methoxybenzotrifluoride; 3-Trifluoromethyl-o-anisidine
Quantity 1g
Health Hazard 2 GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.